This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00833261_g1
See other DBI GE Assays ›
Gene Symbol:
Dbi
Gene Name:
diazepam binding inhibitor
Gene Aliases:
ACBD1, Acbp, EP, endozepine
Chromosome Location:
Chr.1: 120113280 - 120121096 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00833261_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dbi
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001037999.2 NP_001033088.1 2 - 3 278 167
NM_007830.4 NP_031856.1 2 - 3 272 167
GenBank mRNA AA068307.1 - 2 - 3 218 167
AK008540.1 - 2 - 3 193 167
AK008576.1 - 2 - 3 193 167
AK018720.1 - 2 - 3 194 167
AK020692.1 - 2 - 3 242 167
AK027906.1 - 2 - 3 194 167
AK148753.1 - 2 - 3 194 167
AM000002.1 - 2 - 3 278 167
AV001592.1 - 2 - 3 166 167
BC028874.1 - 2 - 3 172 4194069 167
CJ183794.1 - 2 - 3 273 167
EF437365.1 - 2 - 3 179 167
X61431.1 - 2 - 3 191 167

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dbi

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

behavioral fear response
hair follicle development
triglyceride metabolic process
transport
learning or memory
glial cell proliferation
lateral ventricle development
negative regulation of fatty acid beta-oxidation
regulation of synaptic transmission, GABAergic
phosphatidylcholine acyl-chain remodeling
skin development
positive regulation of lipid biosynthetic process
positive regulation of release of sequestered calcium ion into cytosol
long-term synaptic potentiation
fatty-acyl-CoA binding
lipid binding
benzodiazepine receptor binding
long-chain fatty acyl-CoA binding
protein dimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0