This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00834243_g1
See other TDG GE Assays ›
Gene Symbol:
Tdg Gm9855
Gene Name:
thymine DNA glycosylase
thymine DNA glycosylase pseudogene
Gene Aliases:
E130317C12Rik, JZA-3, Jza1
ENSMUSG00000051559
Chromosome Location:
Chr.10: 82629247 - 82650494 on Build GRCm38
Chromosome Location:
Chr.10: 42052558 - 42055453 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
Assay ID Mm00834243_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tdg
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011561.2 NP_035691.2 7 - 8 904 126
NM_172552.3 NP_766140.2 7 - 8 1003 126
GenBank mRNA AF102875.1 - 5 - 6 576 126
AK053870.1 - 7 - 8 931 126
AK164090.1 - 7 - 8 864 126
AK164643.1 - 7 - 8 971 126
AK169768.1 - 7 - 8 927 126
BC010315.1 - 7 - 8 893 3595866 126
BC085470.1 - 7 - 8 950 6416434 126
BC085471.1 - 7 - 8 974 6816716 126
Gene Symbol: Gm9855
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_037190.1 1 900 126
GenBank mRNA AF069519.1 - 1 861 126
AK089915.1 - 1 904 126

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tdg
Click here to find siRNAs for Gene Gm9855

Panther Classification:

Molecular Function -

DNA metabolism protein DNA glycosylase nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
DNA repair
base-excision repair
mismatch repair
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
cellular response to DNA damage stimulus
metabolic process
embryo development
chromatin modification
negative regulation of protein binding
negative regulation of chromatin binding
regulation of gene expression, epigenetic
DNA demethylation
RNA polymerase II transcription cofactor activity
double-stranded DNA binding
protein kinase C binding
protein binding
transcription factor binding
pyrimidine-specific mismatch base pair DNA N-glycosylase activity
hydrolase activity
hydrolase activity, acting on glycosyl bonds
hydrolase activity, hydrolyzing N-glycosyl compounds
DNA N-glycosylase activity
protein domain specific binding
mismatched DNA binding
SUMO binding
protein homodimerization activity
structure-specific DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000007401.mm.1 NM_011561.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000007401.mm.1 NM_011561.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000007401.mm.1 NM_172552.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000007401.mm.1 NM_172552.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000007401.mm.1 NR_037190.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000007401.mm.1 TC1000000928.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline