This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00834622_m1
See other APBB1IP GE Assays ›
Gene Symbol:
Apbb1ip
Gene Name:
amyloid beta (A4) precursor protein-binding, family B, member 1 interacting protein
Gene Aliases:
9930118P07Rik, Prp48
Chromosome Location:
Chr.2: 22755989 - 22875653 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00834622_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Apbb1ip
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019456.2 NP_062329.2 5 - 6 739 64
GenBank mRNA AF020313.1 - 2 - 3 345 64
AK021331.1 - 6 - 7 618 64
AK040084.1 - 5 - 6 675 64
AK041552.1 - 5 - 6 742 64
AK042467.1 - 5 - 6 677 64
AK079736.1 - 5 - 6 973 64
BC003991.1 - 2 - 3 390 64
BC023110.1 - 5 - 6 628 3707478 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000352.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Apbb1ip

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell
signal transduction
positive regulation of cell adhesion
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200001712.mm.1 NM_019456.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200001712.mm.1 NM_019456.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200001712.mm.1 TC0200000352.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200001714.mm.1 NM_019456.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200001714.mm.1 NM_019456.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200001714.mm.1 TC0200000352.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0