This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00836417_g1
See other IDI1 GE Assays ›
Gene Symbol:
Idi1
Gene Name:
isopentenyl-diphosphate delta isomerase
Gene Aliases:
4832416K17Rik
Chromosome Location:
Chr.13: 8885606 - 8892396 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00836417_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Idi1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145360.2 NP_663335.2 2 - 3 564 82
GenBank mRNA AK029302.1 - 2 - 3 197 82
AK084208.1 - 2 - 3 194 82
AK132073.1 - 2 - 3 197 82
AK160832.1 - 2 - 3 198 82
BC004801.1 - 1 - 2 185 3589498 82
BC110313.1 - 2 - 3 202 5343431 82
CT010310.1 - 1 - 2 142 82

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Idi1

Panther Classification:

Molecular Function -

isomerase

Gene Ontology Categories:

Function(s) Process(es)

isoprenoid biosynthetic process
magnesium ion binding
isopentenyl-diphosphate delta-isomerase activity
manganese ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300000307.mm.1 NM_145360.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300000307.mm.1 NM_145360.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300000307.mm.1 TC1300000045.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 565056 - -
ABI Mouse Genome Survey Microarray 826570 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline