This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00838531_g1
See other SWI5 GE Assays ›
Gene Symbol:
Swi5
Gene Name:
SWI5 recombination repair homolog (yeast)
Gene Aliases:
1500019F05Rik, 2900010J23Rik, AI046671
Chromosome Location:
Chr.2: 32278805 - 32288136 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00838531_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Swi5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290552.1 NP_001277481.1 3 - 4 220 146
NM_175190.5 NP_780399.1 3 - 4 365 146
GenBank mRNA AB597964.1 - 3 - 4 213 146
AB597965.1 - 1 - 2 117 146
AK013517.1 - 3 - 4 325 146
AK031907.1 - 3 - 4 333 146
AK041567.1 - 3 - 4 307 146
AK088274.1 - 3 - 4 326 146
AK166608.1 - 3 - 4 309 146
AK216650.1 - 1 - 2 134 146
BC021748.1 - 3 - 4 315 3586619 146

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Swi5

Panther Classification:

Molecular Function -

DNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair via homologous recombination
DNA repair
cellular response to DNA damage stimulus
cellular response to ionizing radiation
protein binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21114429 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21114426 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21114427 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21114428 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21114431 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21114432 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040048 AK166608.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040048 AK216650.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040048 BY683904.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040048 NM_175190.4 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040048 AK013517.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040046 AK166608.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040046 BC021748.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040046 BY683904.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040046 NM_175190.4 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040046 AK013517.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 395111 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928658 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 845280 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 614640 AK089251.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 466635 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 14771 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119937 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040046 AK166608.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040046 BC021748.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040046 BY683904.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040046 NM_175190.4 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040046 AK013517.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040048 AK166608.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040048 AK216650.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040048 BY683904.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040048 NM_175190.4 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040048 AK013517.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21114442 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21114458 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21114440 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21114424 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21114448 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21114419 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21114445 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21114455 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21114438 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21114427 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21114431 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21114453 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21114459 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21114426 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21114451 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21114420 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21114454 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21114456 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21114435 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21114450 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21114429 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21114437 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21114436 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21114433 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21114428 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21114422 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P119937 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00337729 - -
Whole Mouse Genome G4122A A_52_P599181 AK166608.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P599181 AK216650.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P599181 BY683904.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P599181 NM_175190.4 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P599181 AK013517.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P444401 AK039840.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P599175 AK166608.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P599175 AK216650.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P599175 BC021748.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P599175 BY683904.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P599175 NM_175190.4 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135915 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P119937 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01596956 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01596938 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01596943 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01596940 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01596949 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21114431 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21114427 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21114426 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21114428 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21114429 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21114432 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114419 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114351 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114361 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114412 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114387 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114426 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114431 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114400 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114375 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114401 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114429 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114397 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114378 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114394 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114382 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114409 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114415 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114385 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114399 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114363 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114393 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114381 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114371 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114404 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114355 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114390 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114373 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114402 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114422 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114406 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114427 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21114395 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01913558 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02252383 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21114410 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21114415 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21114364 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21114424 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02224977 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21114421 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21114401 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21114413 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21114409 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01596947 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21114420 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21114368 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21114361 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21114404 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21114412 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21114407 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21114402 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21114419 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21114422 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21114363 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21114426 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01312871 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01596943 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21114417 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10481591 - -
Affymetrix Mouse Gene 1.0 ST Array 10481586 - -
Affymetrix Mouse Gene 1.0 ST Array 10481589 - -
Affymetrix Mouse Gene 1.0 ST Array 10481587 - -
Affymetrix Mouse Gene 1.0 ST Array 10481588 - -
Affymetrix HT MG-430 PM Array Plate 1433480_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10481585 - -
Mouse Genome 430 2.0 Array 1433480_at - -
Mouse Genome 430 2.0 Array 1433480_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 72931_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 72931_at - -
Affymetrix Mouse Clariom S Array PSR0200028003.mm.1 NM_175190.5 Refseq
Affymetrix Mouse Clariom S Array PSR0200028003.mm.1 TC0200003299.mm.1 MTA TC
Mouse Expression Set 430 1433480_at - -
Mouse Expression Set 430 1433480_PM_at - -
Murine Genome U74v2 Set 96658_PM_at - -
Murine Genome U74v2 Set 96658_at - -
Mouse Genome 430A 2.0 Array 1433480_PM_at - -
Mouse Genome 430A 2.0 Array 1433480_at - -
Affymetrix HT Mouse Genome 430A Array 1433480_at - -
Affymetrix HT Mouse Genome 430A Array 1433480_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 897158 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 510333 - -
Illumina mouse-6 v1.0 expression beadchip 510333 - -
Illumina mouseRef-8 v1.0 expression beadchip 0000510333 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2733835 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2733835 - -
mouse-6 v1.1 (Illumina) scl19447.7.1_15-S - -
Illumina Sentrix mouse-6.v1 bead array 510333 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl19447.7.1_15-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 510333 - -
MouseRef-8 v2 Expression BeadChip ILMN_2733835 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2733835 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl19447.7.1_15-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jmd5k:80/100.66.78.41:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0