This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00839181_m1
See other RBFOX1 GE Assays ›
Gene Symbol:
Rbfox1
Gene Name:
RNA binding protein, fox-1 homolog (C. elegans) 1
Gene Aliases:
A2bp, A2bp1, Hrnbp1, fox-1
Chromosome Location:
Chr.16: 5884793 - 7412480 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00839181_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbfox1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_183188.2 NP_899011.2 10 - 11 1152 127
GenBank mRNA AF191501.1 - 10 - 11 1153 127
AY659956.1 - 10 - 11 988 127

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600000054.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rbfox1

Panther Classification:

Molecular Function -

RNA metabolism protein RNA processing factor RNA splicing factor nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
RNA splicing
regulation of RNA splicing
neuromuscular process controlling balance
nucleotide binding
nucleic acid binding
RNA binding
protein C-terminus binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600000512.mm.1 NM_183188.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600000512.mm.1 NM_183188.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600000512.mm.1 TC1600000054.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600000513.mm.1 NM_021477.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600000513.mm.1 NM_021477.5 Refseq
Affymetrix Mouse Clariom S Array PSR1600000513.mm.1 NM_183188.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600000513.mm.1 NM_183188.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600000513.mm.1 TC1600000054.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600000517.mm.1 NM_021477.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600000517.mm.1 NM_021477.5 Refseq
Affymetrix Mouse Clariom S Array PSR1600000517.mm.1 NM_183188.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600000517.mm.1 NM_183188.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600000517.mm.1 TC1600000054.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600000518.mm.1 NM_021477.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600000518.mm.1 NM_021477.5 Refseq
Affymetrix Mouse Clariom S Array PSR1600000518.mm.1 NM_183188.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600000518.mm.1 NM_183188.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600000518.mm.1 TC1600000054.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600000515.mm.1 NM_021477.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600000515.mm.1 NM_021477.5 Refseq
Affymetrix Mouse Clariom S Array PSR1600000515.mm.1 NM_183188.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600000515.mm.1 NM_183188.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600000515.mm.1 TC1600000054.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600000516.mm.1 NM_021477.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600000516.mm.1 NM_021477.5 Refseq
Affymetrix Mouse Clariom S Array PSR1600000516.mm.1 NM_183188.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600000516.mm.1 NM_183188.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600000516.mm.1 TC1600000054.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600000519.mm.1 NM_021477.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600000519.mm.1 NM_021477.5 Refseq
Affymetrix Mouse Clariom S Array PSR1600000519.mm.1 NM_183188.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600000519.mm.1 NM_183188.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600000519.mm.1 TC1600000054.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600000520.mm.1 NM_021477.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600000520.mm.1 NM_021477.5 Refseq
Affymetrix Mouse Clariom S Array PSR1600000520.mm.1 NM_183188.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600000520.mm.1 NM_183188.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600000520.mm.1 TC1600000054.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0