This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00839502_m1
See other POLR2A GE Assays ›
Gene Symbol:
Polr2a
Gene Name:
polymerase (RNA) II (DNA directed) polypeptide A
Gene Aliases:
220kDa, Rpb1, Rpo2-1
Chromosome Location:
Chr.11: 69733997 - 69758633 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00839502_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Polr2a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001291068.1 NP_001277997.1 5 - 6 1232 70
GenBank mRNA AK154270.1 - 5 - 6 1234 70
AK154389.1 - 5 - 6 1234 70
AK155391.1 - 5 - 6 1234 70
AK170834.1 - 789 70
U37500.1 - 5 - 6 891 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Endogenous Controls Recommended Assay Mus musculus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of RNA splicing
core promoter binding
RNA polymerase II activity
DNA binding
DNA-directed RNA polymerase activity
protein binding
transferase activity
nucleotidyltransferase activity
ubiquitin protein ligase binding
poly(A) RNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-lsr54:80/100.66.79.61:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0