Assay Details
| Gene Symbol: Fgf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291104.1 | NP_001278033.1 | 2 - 3 | 398 | 98 | |
| NM_010198.2 | NP_034328.1 | 2 - 3 | 398 | 98 | ||
| GenBank mRNA | BC066859.1 | - | 2 - 3 | 398 | 30101881 | 98 |
| U66203.1 | - | 2 - 3 | 307 | 98 | ||
More Information
Set Membership: |
|

Fgf11
Genomic Map