This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00840830_g1
See other MYDGF GE Assays ›
Gene Symbol:
Mydgf
Gene Name:
myeloid derived growth factor
Gene Aliases:
D17Wsu104e, Il25, Ly6elg
Chromosome Location:
Chr.17: 56176541 - 56183920 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00840830_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mydgf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_080837.2 NP_543027.1 4 - 5 373 66
GenBank mRNA AK004074.1 - 4 - 5 373 66
AK008289.1 - 4 - 5 359 66
AK008854.1 - 4 - 5 354 66
AK018016.1 - 4 - 5 356 66
AK133947.1 - 357 66
AK148083.1 - 4 - 5 373 66
AK162834.1 - 4 - 5 361 66
AY038184.1 - 4 - 5 369 66
BC132127.1 - 4 - 5 394 40130404 66
BC132129.1 - 4 - 5 394 40130406 66

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mydgf

Panther Classification:

Molecular Function -

growth factor

Gene Ontology Categories:

Function(s)

angiogenesis
positive regulation of protein phosphorylation
positive regulation of endothelial cell proliferation
apoptotic process
positive regulation of phosphatidylinositol 3-kinase signaling
negative regulation of apoptotic process
positive regulation of MAPK cascade
positive regulation of angiogenesis
positive regulation of transcription from RNA polymerase II promoter
positive regulation of protein kinase B signaling
cellular response to brefeldin A

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline