This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01130951_m1
See other CDH10 GE Assays ›
Gene Symbol:
Cdh10
Gene Name:
cadherin 10
Gene Aliases:
A830016G23Rik, C030003B10Rik, C030011H18Rik
Chromosome Location:
Chr.15: 18818888 - 19016206 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01130951_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdh10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009865.2 NP_033995.1 7 - 8 1739 77
GenBank mRNA AF183946.1 - 7 - 8 1508 77
AK138606.1 - 7 - 8 1741 77
BC052056.1 - 7 - 8 1447 5696194 77
BC062962.1 - 7 - 8 1447 5696194 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500000125.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
calcium ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500001113.mm.1 NM_009865.3 Refseq
Affymetrix Mouse Clariom S Array PSR1500001113.mm.1 TC1500000125.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0