This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01132037_m1
See other SUGCT GE Assays ›
Gene Symbol:
Sugct
Gene Name:
succinyl-CoA glutarate-CoA transferase
Gene Aliases:
5033411D12Rik, D17907
Chromosome Location:
Chr.13: 16857472 - 17694793 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01132037_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sugct
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_138654.3 NP_619595.3 8 - 9 753 125
GenBank mRNA AA108199.1 - 2 - 3 205 125
AF397014.1 - 8 - 9 769 125
AK160265.1 - 7 - 8 626 125
BC055852.1 - 8 - 9 727 4221852 125

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sugct

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
catalytic activity
transferase activity
succinate-hydroxymethylglutarate CoA-transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0