This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01132874_m1
See other FAM173B GE Assays ›
Gene Symbol:
Fam173b
Gene Name:
family with sequence similarity 173, member B
Gene Aliases:
A930016P21Rik, AA987072
Chromosome Location:
Chr.15: 31601941 - 31617537 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01132874_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fam173b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026546.3 NP_080822.1 4 - 5 505 77
GenBank mRNA AK020868.1 - 4 - 5 505 77
AK160923.1 - 632 77
BC030179.1 - 3 - 4 496 5039149 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500000208.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fam173b

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30115250 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P117966 AK160923.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P117966 BC030179.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P117966 AK020868.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P117966 AK044518.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P117966 NM_026546.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P117966 AK160923.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P117966 BC030179.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P117966 AK020868.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P117966 AK044518.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P117966 NM_026546.3 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 743004 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 757879 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 46826 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 561605 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 961938 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 544558 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 788461 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 818067 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 440684 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 720516 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 52477 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 141549 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 37170 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154556 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161592 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P117966 AK160923.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P117966 BC030179.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P117966 AK020868.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P117966 AK044518.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P117966 NM_026546.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P117966 AK160923.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P117966 BC030179.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P117966 AK020868.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P117966 AK044518.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P117966 NM_026546.3 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115266 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115272 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115256 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115254 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115251 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115269 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115255 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115259 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115270 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115258 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115262 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115264 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30115261 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154556 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161592 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08326072 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08326063 - -
Whole Mouse Genome G4122A A_52_P502424 AK044518.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P117966 AK160923.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P117966 BC030179.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P117966 AK020868.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P117966 AK044518.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P117966 NM_026546.3 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P117966 AK160923.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P117966 BC030179.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P117966 AK020868.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P117966 AK044518.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P117966 NM_026546.3 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112781 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120340 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154556 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08326072 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161592 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12140218 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12140217 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30115250 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115333 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115285 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115358 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115357 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115334 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115297 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115372 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115296 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115337 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115293 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115277 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115304 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115264 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115384 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115335 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115386 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115323 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115319 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115340 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115286 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115310 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115343 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115247 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115376 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115354 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115308 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115271 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115371 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115256 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115289 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115329 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115347 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115345 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115254 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115378 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115330 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115291 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115300 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115273 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115388 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115351 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115248 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115284 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115355 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115364 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115368 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115288 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115324 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115267 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115278 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115339 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115316 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115379 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115325 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115269 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115366 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115342 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115294 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115299 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115349 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115375 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115341 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30115326 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115254 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115271 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115264 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12008809 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11916538 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115368 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12451714 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12399986 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115366 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115337 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115371 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12008816 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115351 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115388 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12070105 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11855650 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115283 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115277 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12062763 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115361 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115274 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115343 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12141399 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115256 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11892087 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12035980 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115372 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115358 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115386 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115270 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115373 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115259 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12070107 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115364 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12233129 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115285 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115269 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11855649 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115262 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115278 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115291 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115266 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12141406 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12140218 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12131378 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115248 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115258 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115347 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12166840 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115313 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115339 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115260 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115303 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12008817 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12056207 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115275 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115318 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115316 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11912140 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115355 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11892086 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115325 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115272 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115261 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115284 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115341 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12358964 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115299 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115349 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115288 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30115300 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10423515 - -
Affymetrix Mouse Gene 1.0 ST Array 10423516 - -
Affymetrix Mouse Gene 1.0 ST Array 10423514 - -
Affymetrix Mouse Gene 1.0 ST Array 10423517 - -
Affymetrix Mouse Gene 1.0 ST Array 10423513 - -
Affymetrix HT MG-430 PM Array Plate 1444686_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1428683_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10423512 - -
Mouse Genome 430 2.0 Array 1457848_at - -
Mouse Genome 430 2.0 Array 1428683_PM_at - -
Mouse Genome 430 2.0 Array 1444686_PM_at - -
Mouse Genome 430 2.0 Array 1444686_at - -
Mouse Genome 430 2.0 Array 1457848_PM_at - -
Mouse Genome 430 2.0 Array 1428683_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 68073_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 68073_PM_at - -
Affymetrix Mouse Clariom S Array PSR1500001546.mm.1 NM_026546.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500001546.mm.1 NM_026546.3 Refseq
Affymetrix Mouse Clariom S Array PSR1500001546.mm.1 TC1500000208.mm.1 MTA TC
Mouse Expression Set 430 1428683_PM_at - -
Mouse Expression Set 430 1428683_at - -
Mouse Expression Set 430 1444686_PM_at - -
Mouse Expression Set 430 1444686_at - -
Affymetrix HT Mouse Genome 430B Array 1428683_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1444686_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1428683_at - -
Affymetrix HT Mouse Genome 430B Array 1444686_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 886949 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 105290520 - -
Illumina mouse-6 v1.0 expression beadchip 105290520 - -
Illumina mouse-6 v1.0 expression beadchip 670546 - -
Illumina mouseRef-8 v1.0 expression beadchip 0000670546 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1224980 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1224980 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2500032 - -
mouse-6 v1.1 (Illumina) scl48019.6_564-S - -
mouse-6 v1.1 (Illumina) scl0068073.2_190-S - -
Illumina Sentrix mouse-6.v1 bead array 670546 - -
Illumina Sentrix mouse-6.v1 bead array 105290520 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0068073.2_190-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 105290520 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 670546 - -
MouseRef-8 v2 Expression BeadChip ILMN_1224980 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1224980 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0068073.2_190-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline