This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01134787_g1
See other AGER GE Assays ›
Gene Symbol:
Ager
Gene Name:
advanced glycosylation end product-specific receptor
Gene Aliases:
RAGE
Chromosome Location:
Chr.17: 34597410 - 34600937 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01134787_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ager
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271422.1 NP_001258351.1 4 - 5 468 72
NM_001271423.1 NP_001258352.1 4 - 5 441 72
NM_001271424.1 NP_001258353.1 3 - 4 563 72
NM_007425.3 NP_031451.2 4 - 5 468 72
NR_073173.1 4 - 5 468 72
NR_073175.1 3 - 4 663 72
NR_073176.1 3 - 4 663 72
GenBank mRNA AB207883.1 - 4 - 5 428 72
AK004802.1 - 4 - 5 456 72
BC061182.1 - 4 - 5 456 30300427 72
EU520325.1 - 4 - 5 424 72
EU570240.1 - 4 - 5 424 72
EU570241.1 - 3 - 4 619 72
EU570242.1 - 4 - 5 424 72
EU570243.1 - 4 - 5 424 72
EU570244.1 - 3 - 4 619 72
EU570245.1 - 3 - 4 619 72
EU570246.1 - 3 - 4 581 72
EU570247.1 - 3 - 4 619 72
EU906862.1 - 4 - 5 424 72
EU906863.1 - 4 - 5 397 72
EU906864.1 - 3 - 4 619 72
GU164727.1 - 3 - 4 538 72
GU164728.1 - 4 - 5 443 72
GU164730.1 - 4 - 5 443 72
GU164731.1 - 4 - 5 443 72
GU164732.1 - 4 - 5 443 72
KC692918.1 - 4 - 5 424 72
L33412.1 - 4 - 5 429 72

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ager

Panther Classification:

Molecular Function -

cell adhesion molecule immunoglobulin superfamily cell adhesion molecule

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
negative regulation of protein phosphorylation
positive regulation of protein phosphorylation
negative regulation of endothelial cell proliferation
inflammatory response
negative regulation of cell adhesion
JAK-STAT cascade
positive regulation of autophagy
negative regulation of endothelial cell migration
positive regulation of gene expression
positive regulation of epithelial to mesenchymal transition
positive regulation of fibroblast migration
positive regulation of smooth muscle cell migration
positive regulation of cell migration
neuron projection development
negative regulation of collagen biosynthetic process
positive regulation of apoptotic process
positive regulation of JUN kinase activity
positive regulation of neuron apoptotic process
positive regulation of fibroblast proliferation
positive regulation of smooth muscle cell proliferation
regulation of inflammatory response
positive regulation of inflammatory response
induction of positive chemotaxis
positive regulation of NF-kappaB transcription factor activity
regulation of DNA binding
calcium ion homeostasis
positive regulation of phagocytosis, engulfment
protein localization to membrane
positive regulation of potassium ion transmembrane transporter activity
positive regulation of neuron death
positive regulation of endothelial cell apoptotic process
positive regulation of reactive oxygen species metabolic process
positive regulation of type B pancreatic cell apoptotic process
protein binding
identical protein binding
S100 protein binding
advanced glycation end-product receptor activity
high mobility group box 1 binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0