This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01134874_g1
See other ASIC3 GE Assays ›
Gene Symbol:
Asic3
Gene Name:
acid-sensing (proton-gated) ion channel 3
Gene Aliases:
AW742291, Accn3, DRASIC, SLNAC1, TNAC1
Chromosome Location:
Chr.5: 24410289 - 24417834 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01134874_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Asic3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_183000.2 NP_892045.2 2 - 3 950 74
GenBank mRNA AY261387.1 - 2 - 3 689 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000214.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Asic3

Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
cation transport
sodium ion transport
response to heat
response to mechanical stimulus
response to acidic pH
ion transmembrane transport
sodium ion transmembrane transport
enterobactin transport
detection of chemical stimulus involved in sensory perception
sensory perception of sour taste
detection of temperature stimulus involved in sensory perception
detection of temperature stimulus involved in sensory perception of pain
detection of mechanical stimulus involved in sensory perception of pain
detection of chemical stimulus involved in sensory perception of pain
detection of mechanical stimulus involved in sensory perception
cation transmembrane transport
cation channel activity
sodium channel activity
ligand-gated sodium channel activity
PDZ domain binding
enterobactin transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500001586.mm.1 TC0500000214.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500001586.mm.1 NM_183000.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500001586.mm.1 NM_183000.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0