This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01134932_g1
See other ABCB8 GE Assays ›
Gene Symbol:
Abcb8
Gene Name:
ATP-binding cassette, sub-family B (MDR/TAP), member 8
Gene Aliases:
4833412N02Rik, AA409895
Chromosome Location:
Chr.5: 24393681 - 24409947 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01134932_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Abcb8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029020.2 NP_083296.2 2 - 3 745 111
GenBank mRNA AK014319.1 - 2 - 3 476 111
AK014688.1 - 2 - 3 498 111
AK030624.1 - 2 - 3 472 111
AK038760.1 - 2 - 3 472 111
AK049152.1 - 2 - 3 489 111
AK076315.1 - 2 - 3 496 111
AK166704.1 - 2 - 3 479 111
BC015301.1 - 2 - 3 745 4237894 111

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000213.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Abcb8

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
transmembrane transport
nucleotide binding
ATP binding
ATPase activity
ATPase activity, coupled to transmembrane movement of substances

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500001558.mm.1 TC0500000213.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500001558.mm.1 NM_029020.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500001558.mm.1 NM_029020.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0