This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01135629_g1
See other IMPDH1 GE Assays ›
Gene Symbol:
Impdh1
Gene Name:
inosine 5'-phosphate dehydrogenase 1
Gene Aliases:
B930086D20Rik, IMPDH-I
Chromosome Location:
Chr.6: 29200437 - 29216364 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01135629_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Impdh1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK047537.1 - 4 - 5 531 90
AK089383.1 - 4 - 5 495 90
AK171139.1 - 4 - 5 506 90
AK199145.1 - 1 - 2 51 90
BC053416.1 - 3 - 4 355 6533681 90
CK619435.1 - 4 - 5 505 90
U00978.1 - 3 - 4 250 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600001974.mm.1

Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600015510.mm.1 TC0600001974.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600015510.mm.1 NM_011829.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w64qk:80/100.66.78.26:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0