This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01136817_m1
See other ARHGAP6 GE Assays ›
Gene Symbol:
Arhgap6
Gene Name:
Rho GTPase activating protein 6
Gene Aliases:
AI504284, RhoGAPX-1
Chromosome Location:
Chr.X: 168795094 - 169304440 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01136817_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arhgap6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001287530.1 NP_001274459.1 7 - 8 1243 69
NM_009707.4 NP_033837.2 7 - 8 2611 69
NM_178754.3 NP_848869.1 7 - 8 1154 69
NR_109847.1 7 - 8 1154 69
GenBank mRNA AF012273.2 - 1733 69
AF177664.1 - 1733 69
AK048162.1 - 7 - 8 1108 69
AK048507.1 - 7 - 8 1109 69
AK077996.1 - 7 - 8 1116 69
AK079651.1 - 7 - 8 1067 69
AK166015.1 - 7 - 8 1074 69
AK170575.1 - 7 - 8 1110 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00001700.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Arhgap6

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

actin filament organization
signal transduction
activation of phospholipase C activity
positive regulation of phospholipase activity
regulation of GTPase activity
positive regulation of GTPase activity
focal adhesion assembly
negative regulation of stress fiber assembly
negative regulation of focal adhesion assembly
GTPase activator activity
phospholipase activator activity
SH3 domain binding
phospholipase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00010866.mm.1 NM_001287530.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00010866.mm.1 NM_009707.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00010866.mm.1 NM_178754.3 Refseq
Affymetrix Mouse Clariom S Array PSR0X00010866.mm.1 NR_109847.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00010866.mm.1 TC0X00001700.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00010863.mm.1 NM_001287530.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00010863.mm.1 NM_009707.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00010863.mm.1 NM_178754.3 Refseq
Affymetrix Mouse Clariom S Array PSR0X00010863.mm.1 NR_109847.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00010863.mm.1 TC0X00001700.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rr9bx:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0