This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01137817_g1
See other PPP4C GE Assays ›
Gene Symbol:
Ppp4c
Gene Name:
protein phosphatase 4, catalytic subunit
Gene Aliases:
1110002D08Rik, AU016079, Ppx
Chromosome Location:
Chr.7: 126785866 - 126792471 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01137817_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp4c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019674.3 NP_062648.1 4 - 5 321 87
GenBank mRNA AF088911.1 - 4 - 5 346 87
AK027923.1 - 2 - 3 100 87
AK144244.1 - 3 - 4 248 87
BC001993.1 - 4 - 5 321 3491568 87

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700004295.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp4c

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

regulation of double-strand break repair via homologous recombination
dephosphorylation
signal transduction by protein phosphorylation
NIK/NF-kappaB signaling
NF-kappaB-inducing kinase activity
phosphoprotein phosphatase activity
protein serine/threonine phosphatase activity
protein binding
lamin binding
hydrolase activity
phosphatase activity
protein complex binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700035938.mm.1 NM_019674.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700035938.mm.1 NM_019674.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700035938.mm.1 TC0700004295.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700035937.mm.1 NM_019674.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700035937.mm.1 NM_019674.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700035937.mm.1 TC0700004295.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0