This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01137897_m1
See other PTPRE GE Assays ›
Gene Symbol:
Ptpre
Gene Name:
protein tyrosine phosphatase, receptor type, E
Gene Aliases:
PTPe, PTPepsilon, RPTPepsilon
Chromosome Location:
Chr.7: 135537645 - 135686294 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01137897_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ptpre
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011212.3 NP_035342.3 11 - 12 1168 83
GenBank mRNA AK132455.1 - 10 - 11 1131 83
AK154910.1 - 8 - 9 720 83
AK160947.1 - 8 - 9 717 83
BC053408.1 - 11 - 12 944 83
BC098473.1 - 11 - 12 932 83
D83484.1 - 11 - 12 1505 83
U35368.1 - 11 - 12 1168 83
U36758.1 - 8 - 9 717 83
U40280.1 - 10 - 11 908 83
U62387.1 - 11 - 12 1070 83
Z23052.1 - 2 - 3 176 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700001959.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ptpre

Panther Classification:

Molecular Function -

protein phosphatase protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
transmembrane receptor protein tyrosine phosphatase signaling pathway
dephosphorylation
regulation of mast cell activation
peptidyl-tyrosine dephosphorylation
negative regulation of insulin receptor signaling pathway
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
hydrolase activity
phosphatase activity
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700017210.mm.1 NM_011212.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700017210.mm.1 NM_011212.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700017210.mm.1 TC0700001959.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline