This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01138034_m1
See other ATPAF2 GE Assays ›
Gene Symbol:
Atpaf2
Gene Name:
ATP synthase mitochondrial F1 complex assembly factor 2
Gene Aliases:
ATP12, ATP12p
Chromosome Location:
Chr.11: 60400624 - 60418791 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01138034_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atpaf2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145427.2 NP_663402.2 4 - 5 638 85
GenBank mRNA AK078500.1 - 4 - 5 583 85
BC013607.1 - 4 - 5 544 3585582 85

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100002874.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atpaf2

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

biological_process
proton-transporting ATP synthase complex assembly
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100026141.mm.1 NM_145427.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100026141.mm.1 NM_145427.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100026141.mm.1 TC1100002874.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-ln7zh:80/100.66.79.20:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0