This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01138521_g1
See other GPSM3 GE Assays ›
Gene Symbol:
Gpsm3
Gene Name:
G-protein signalling modulator 3 (AGS3-like, C. elegans)
Gene Aliases:
AA960287, AGS4
Chromosome Location:
Chr.17: 34589806 - 34591754 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01138521_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpsm3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_134116.5 NP_598877.1 2 - 3 553 61
GenBank mRNA AK144833.1 - 2 - 3 535 61
AK146245.1 - 2 - 3 544 61
AK155617.1 - 2 - 3 554 61
BC021942.1 - 2 - 3 143 5068831 61
BC080804.1 - 3 - 4 256 5355970 61
BC085186.1 - 2 - 3 535 30680693 61

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpsm3

Gene Ontology Categories:

Function(s) Process(es)

regulation of G-protein coupled receptor protein signaling pathway
GDP-dissociation inhibitor activity
GTPase regulator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700006164.mm.1 NM_134116.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700006164.mm.1 NM_134116.5 Refseq
Affymetrix Mouse Clariom S Array PSR1700006164.mm.1 TC1700002783.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700006163.mm.1 NM_134116.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700006163.mm.1 NM_134116.5 Refseq
Affymetrix Mouse Clariom S Array PSR1700006163.mm.1 TC1700002783.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2tqsm:80/100.66.78.74:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0