This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01138522_g1
See other GPSM3 GE Assays ›
Gene Symbol:
Gpsm3
Gene Name:
G-protein signalling modulator 3 (AGS3-like, C. elegans)
Gene Aliases:
AA960287, AGS4
Chromosome Location:
Chr.17: 34589806 - 34591754 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01138522_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpsm3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_134116.5 NP_598877.1 3 - 4 758 63
GenBank mRNA AK146245.1 - 3 - 4 749 63
AK155863.1 - 3 - 4 436 63
BC021942.1 - 3 - 4 348 5068831 63
BC080804.1 - 4 - 5 461 5355970 63
BC085186.1 - 3 - 4 740 30680693 63
BQ031296.1 - 1 - 2 63

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpsm3

Gene Ontology Categories:

Function(s) Process(es)

regulation of G-protein coupled receptor protein signaling pathway
GDP-dissociation inhibitor activity
GTPase regulator activity

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957633 BC080804.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957633 BC085186.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957633 BQ031296.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957633 AK155617.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957633 AK144833.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 798436 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P103682 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957633 BC080804.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957633 BC085186.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957633 BQ031296.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957633 AK155617.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957633 AK144833.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154632 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154637 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154646 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154625 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154628 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154623 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154649 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154643 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154633 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154642 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154630 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154641 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154627 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154638 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154645 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154634 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154640 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154647 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154631 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154635 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154622 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P103682 - -
Whole Mouse Genome G4122A A_51_P106322 BC080804.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P106322 BC085186.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P106322 BQ031296.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P106322 AK155617.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P106322 AK155863.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108458 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P103682 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13352426 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13352432 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13352428 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13352429 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13352440 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154724 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154694 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154633 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154656 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154642 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154679 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154675 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154674 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154628 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154696 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154649 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154720 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154623 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154637 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154672 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154666 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154638 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154645 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154719 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154631 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154715 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154684 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154704 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154700 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154652 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154653 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154717 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154668 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154662 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154702 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154676 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154710 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154706 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154708 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154688 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154647 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154686 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154665 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154646 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154723 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154669 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154622 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13352429 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154628 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154646 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154634 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154643 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154649 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154630 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13214273 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154645 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154637 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154638 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154625 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154623 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154633 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154642 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154641 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31154627 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10444391 - -
Affymetrix Mouse Gene 1.0 ST Array 10444387 - -
Affymetrix Mouse Gene 1.0 ST Array 10444389 - -
Affymetrix Mouse Gene 1.0 ST Array 10444393 - -
Affymetrix Mouse Gene 1.0 ST Array 10444390 - -
Affymetrix Mouse Gene 1.0 ST Array 10444388 - -
Affymetrix Mouse Gene 1.0 ST Array 10444392 - -
Affymetrix HT MG-430 PM Array Plate 1418396_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10444386 - -
Mouse Genome 430 2.0 Array 1418396_at - -
Mouse Genome 430 2.0 Array 1418396_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 106512_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 106512_at - -
Affymetrix Mouse Clariom S Array PSR1700024750.mm.1 NM_134116.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024750.mm.1 NM_134116.5 Refseq
Affymetrix Mouse Clariom S Array PSR1700024750.mm.1 TC1700002783.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700006168.mm.1 NM_134116.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700006168.mm.1 NM_134116.5 Refseq
Affymetrix Mouse Clariom S Array PSR1700006168.mm.1 TC1700002783.mm.1 MTA TC
Mouse Expression Set 430 1418396_at - -
Mouse Expression Set 430 1418396_PM_at - -
Murine Genome U74v2 Set 94442_s_at - -
Murine Genome U74v2 Set 94441_PM_at - -
Murine Genome U74v2 Set 94442_PM_s_at - -
Murine Genome U74v2 Set 94441_at - -
Mouse Genome 430A 2.0 Array 1418396_at - -
Mouse Genome 430A 2.0 Array 1418396_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1418396_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1418396_at - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 2690154 - -
Illumina mouseRef-8 v1.0 expression beadchip 0002690154 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1222256 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1222256 - -
mouse-6 v1.1 (Illumina) scl50818.3.28_5-S - -
Illumina Sentrix mouse-6.v1 bead array 2690154 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl50818.3.28_5-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 2690154 - -
MouseRef-8 v2 Expression BeadChip ILMN_1222256 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1222256 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0