This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01139534_m1
See other FBXL13 GE Assays ›
Gene Symbol:
Fbxl13
Gene Name:
F-box and leucine-rich repeat protein 13
Gene Aliases:
4921539K22Rik
Chromosome Location:
Chr.5: 21483847 - 21645698 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01139534_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fbxl13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001199632.1 NP_001186561.1 8 - 9 911 89
NM_177076.4 NP_796050.2 8 - 9 911 89
GenBank mRNA AK029566.1 - 8 - 9 913 89
AK030172.1 - 8 - 9 894 89
BC145291.1 - 8 - 9 837 9053845 89
BC145292.1 - 8 - 9 838 9053846 89

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fbxl13

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline