This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01144399_g1
See other SYNCRIP GE Assays ›
Gene Symbol:
Syncrip
Gene Name:
synaptotagmin binding, cytoplasmic RNA interacting protein
Gene Aliases:
2610109K23Rik, 4632417O19Rik, GRY-RBP, Nsap1, Nsap1l, pp68
Chromosome Location:
Chr.9: 88449364 - 88482592 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01144399_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Syncrip
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001284328.1 NP_001271257.1 8 - 9 1359 79
NM_019666.2 NP_062640.2 8 - 9 1359 79
NM_019796.5 NP_062770.1 8 - 9 1359 79
GenBank mRNA AB035725.1 - 8 - 9 1158 79
AF093821.1 - 7 - 8 1020 79
AF408434.1 - 6 - 7 812 79
AK034845.1 - 8 - 9 1171 79
AK076026.1 - 8 - 9 1149 79
AK077588.1 - 8 - 9 1108 79
AK078158.1 - 6 - 7 792 79
AK083398.1 - 7 - 8 950 79
AK162448.1 - 2 - 3 241 79
BC041148.1 - 8 - 9 1204 4946652 79
BC050079.1 - 8 - 9 1132 30012104 79
BC055863.1 - 8 - 9 1137 79
BC058807.1 - 8 - 9 1226 79
BC080309.1 - 8 - 9 1062 5716184 79
BC108363.1 - 8 - 9 1093 4239809 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900003342.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Syncrip

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

osteoblast differentiation
mRNA processing
RNA splicing
negative regulation of translation
CRD-mediated mRNA stabilization
cellular response to interferon-gamma
nucleotide binding
nucleic acid binding
RNA binding
poly(A) binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900023218.mm.1 NM_001284328.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900023218.mm.1 NM_019666.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900023218.mm.1 NM_019666.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900023218.mm.1 NM_019796.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900023218.mm.1 TC0900003342.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900023214.mm.1 NM_001284328.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900023214.mm.1 NM_019666.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900023214.mm.1 NM_019666.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900023214.mm.1 NM_019796.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900023214.mm.1 TC0900003342.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900023215.mm.1 NM_001284328.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900023215.mm.1 NM_019666.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900023215.mm.1 NM_019666.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900023215.mm.1 NM_019796.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900023215.mm.1 TC0900003342.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900023219.mm.1 NM_001284328.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900023219.mm.1 NM_019666.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900023219.mm.1 NM_019666.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900023219.mm.1 NM_019796.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900023219.mm.1 TC0900003342.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900023216.mm.1 NM_001284328.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900023216.mm.1 NM_019666.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900023216.mm.1 NM_019666.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900023216.mm.1 NM_019796.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900023216.mm.1 TC0900003342.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0