This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01144796_m1
See other PRSS57 GE Assays ›
Gene Symbol:
Prss57
Gene Name:
protease, serine 57
Gene Aliases:
2900092M14Rik, GLGL782, Prssl1, UNQ782
Chromosome Location:
Chr.10: 79781474 - 79795605 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01144796_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prss57
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042710.1 NP_001036175.1 3 - 4 445 90
GenBank mRNA AK158376.1 - 3 - 4 445 91
BC116380.2 - 4 - 5 595 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

protease serine protease protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27278965 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136426 BC061052.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136426 AK082898.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136426 AK087247.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136426 AK079286.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136426 NM_031380.2 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 165276 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 681084 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 751944 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 582088 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777116 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 142966 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 48214 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136426 BC061052.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136426 AK082898.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136426 AK087247.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136426 AK079286.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136426 NM_031380.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P315820 AK158376.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P315820 NM_001042710.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P315820 AK013841.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P315820 AK153740.1 mouse_transcriptome
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08305244 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08305245 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08305247 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08305250 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27278965 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278914 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278845 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278857 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278871 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278913 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278841 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278872 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278974 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278910 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278971 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278976 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278990 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278897 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278984 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278852 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278919 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278884 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278983 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278902 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278928 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278955 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278949 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278923 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278839 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278882 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278908 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278936 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278887 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278960 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278926 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278874 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278967 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278947 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278987 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278945 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278846 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278978 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278867 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278876 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278934 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278924 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278918 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278980 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278995 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278899 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278941 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278943 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278847 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278895 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278850 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278964 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278859 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278907 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278965 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278878 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278938 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278893 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278915 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278917 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278979 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278861 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278866 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278961 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278963 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278973 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278905 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278892 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278929 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278854 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27278917 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27278915 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08485565 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27278921 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08241880 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27278936 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08920333 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08561866 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08625288 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27278931 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27278952 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08920338 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27278945 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27278942 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08920337 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27278926 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27278924 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27278934 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27278928 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27278914 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27278935 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27278920 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08924994 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27278919 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27278938 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08677736 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27278941 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27278947 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27278949 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08241879 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10364501 - -
Affymetrix Mouse Gene 1.0 ST Array 10370646 - -
Affymetrix Mouse Gene 1.0 ST Array 10370650 - -
Affymetrix Mouse Gene 1.0 ST Array 10364500 - -
Affymetrix Mouse Gene 1.0 ST Array 10370647 - -
Affymetrix Mouse Gene 1.0 ST Array 10370649 - -
Affymetrix Mouse Gene 1.0 ST Array 10370645 - -
Affymetrix Mouse Gene 1.0 ST Array 10370648 - -
Affymetrix Mouse Gene 1.1 ST Array 10370644 - -
Affymetrix Mouse Gene 1.1 ST Array 10364494 - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 299800 - -
Array Name Probe IDs Targets Target Type
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1218587 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1218587 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0