This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01144799_m1
See other ATP5D GE Assays ›
Gene Symbol:
Atp5d
Gene Name:
ATP synthase, H+ transporting, mitochondrial F1 complex, delta subunit
Gene Aliases:
0610008F14Rik, 1500000I11Rik, AA960090, AI876556, AU020773, C85518
Chromosome Location:
Chr.10: 80142280 - 80145818 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01144799_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp5d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025313.2 NP_079589.2 2 - 3 451 60
GenBank mRNA AK002330.1 - 2 - 3 401 60
AK005082.1 - 2 - 3 395 60
AK018011.1 - 2 - 3 397 60
AK076064.1 - 2 - 3 376 60
AK151151.1 - 2 - 3 386 60
AK155285.1 - 2 - 3 435 60
AK165932.1 - 2 - 3 400 60
BC008273.1 - 2 - 3 360 2811419 60
CT010177.1 - 2 - 3 300 60

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000003218.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atp5d

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

ATP biosynthetic process
transport
ion transport
ATP synthesis coupled proton transport
proton transport
ATP metabolic process
hydrogen ion transmembrane transport
hydrogen ion transmembrane transporter activity
ATPase activity
protein complex binding
proton-transporting ATP synthase activity, rotational mechanism
proton-transporting ATPase activity, rotational mechanism

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000006289.mm.1 NM_025313.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000006289.mm.1 NM_025313.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000006289.mm.1 TC1000003218.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0