This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01144803_g1
See other CIRBP GE Assays ›
Gene Symbol:
Cirbp
Gene Name:
cold inducible RNA binding protein
Gene Aliases:
Cirp, R74941
Chromosome Location:
Chr.10: 80165990 - 80172691 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01144803_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cirbp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007705.2 NP_031731.1 5 - 6 517 98
GenBank mRNA AK014655.1 - 5 - 6 520 98
AK132584.1 - 5 - 6 520 98
BC075699.1 - 5 - 6 507 30656047 98
D78135.1 - 5 - 6 517 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000843.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cirbp

Gene Ontology Categories:

Function(s) Process(es)

response to UV
negative regulation of translation
stress granule assembly
positive regulation of translation
mRNA stabilization
nucleotide binding
nucleic acid binding
RNA binding
mRNA 3'-UTR binding
translation repressor activity
poly(A) RNA binding
small ribosomal subunit rRNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000006335.mm.1 NM_007705.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000006335.mm.1 NM_007705.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000006335.mm.1 TC1000000843.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000006333.mm.1 NM_007705.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000006333.mm.1 NM_007705.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000006333.mm.1 TC1000000843.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0