This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01145447_m1
See other ILDR1 GE Assays ›
Gene Symbol:
Ildr1
Gene Name:
immunoglobulin-like domain containing receptor 1
Gene Aliases:
AU041483, AU044638
Chromosome Location:
Chr.16: 36693978 - 36726804 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01145447_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ildr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001285788.1 NP_001272717.1 2 - 3 403 79
NM_001285791.1 NP_001272720.1 2 - 3 403 79
NM_134109.2 NP_598870.1 2 - 3 403 79
GenBank mRNA AK035504.1 - 2 - 3 79
AK036844.1 - 2 - 3 329 79
AK050806.1 - 2 - 3 349 79
AK136284.1 - 2 - 3 403 79
BC010485.1 - 2 - 3 343 3483341 79
BC057644.1 - 2 - 3 400 4912789 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600000522.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ildr1

Panther Classification:

Molecular Function -

defense/immunity protein immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

receptor-mediated endocytosis
positive regulation of peptide hormone secretion
high-density lipoprotein particle receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600004885.mm.1 NM_001285788.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600004885.mm.1 NM_001285791.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600004885.mm.1 NM_134109.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600004885.mm.1 TC1600000522.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rr9bx:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0