This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01146238_m1
See other CCT8 GE Assays ›
Gene Symbol:
Cct8
Gene Name:
chaperonin containing Tcp1, subunit 8 (theta)
Gene Aliases:
AI132397, Cctq, Tcpq
Chromosome Location:
Chr.16: 87483325 - 87495869 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01146238_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cct8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009840.3 NP_033970.3 3 - 4 315 75
GenBank mRNA AK010882.1 - 3 - 4 316 75
AK075711.1 - 3 - 4 296 75
AK075880.1 - 3 - 4 304 75
AK145661.1 - 3 - 4 299 75
AK145754.1 - 3 - 4 298 75
AK145918.1 - 3 - 4 317 75
AK147038.1 - 3 - 4 318 75
AK159969.1 - 3 - 4 307 75
AK168465.1 - 3 - 4 287 75
AK172867.1 - 3 - 4 286 75
AK184650.1 - 2 - 3 115 75
AK189477.1 - 2 - 3 168 75
BC009007.1 - 3 - 4 273 3582639 75
Z37164.1 - 3 - 4 319 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600001986.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cct8

Panther Classification:

Molecular Function -

chaperonin

Gene Ontology Categories:

Function(s) Process(es)

protein folding
binding of sperm to zona pellucida
pore complex assembly
toxin transport
nucleotide binding
protein binding
ATP binding
unfolded protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600015445.mm.1 NM_009840.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600015445.mm.1 NM_009840.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600015445.mm.1 TC1600001986.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0