This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01146716_m1
See other NXPE3 GE Assays ›
Gene Symbol:
Nxpe3
Gene Name:
neurexophilin and PC-esterase domain family, member 3
Gene Aliases:
Fam55c, Gm1752, Gm611
Chromosome Location:
Chr.16: 55839953 - 55896773 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01146716_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nxpe3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001134457.1 NP_001127929.1 5 - 6 1648 106
GenBank mRNA AK157499.1 - 5 - 6 1655 106
BC150961.1 - 4 - 5 1246 9087872 106
BC150969.1 - 4 - 5 1246 9087882 106

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600001743.mm.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963364 AK157499.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963364 AK034283.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963364 NM_001134494.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963364 NM_001134457.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P690909 AK034283.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P690909 NM_001134457.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P690909 AK034283.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P690909 NM_001134457.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 717136 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 549382 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 268150 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 81745 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 746754 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 664238 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 358990 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 499659 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 849553 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 865481 AK013591.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 865481 BC066016.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 865481 XM_001481319.1 refseq_mrna_mouse_XM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 865481 XM_129042.8 refseq_mrna_mouse_XM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 2361 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 692841 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 189449 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 812806 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 915808 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 121176 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 326797 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 613929 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 396061 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 866453 AK035463.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 754517 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 420084 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 799676 AF371320.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 799676 AK013358.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 799676 AK079337.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 799676 AK079550.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 799676 BC150157.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 799676 BC150479.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 799676 NM_080640.4 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 849217 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 29856 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 891252 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 343981 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 252545 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 125137 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 46224 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 65619 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 163571 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 89628 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 67930 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 5335 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P143338 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963364 AK157499.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963364 AK034283.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963364 NM_001134494.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963364 NM_001134457.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P690909 AK034283.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P690909 NM_001134457.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P690909 AK034283.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P690909 NM_001134457.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P143338 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08606920 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08606948 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08606899 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08606910 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03200773 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08606969 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08606940 - -
Whole Mouse Genome G4122A A_52_P690909 AK034283.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P690909 NM_001134457.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P690909 AK034283.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P690909 NM_001134457.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P579411 BC150969.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P579411 AK157499.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P579411 BC150961.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P579411 NM_001134494.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P579411 NM_001134457.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114712 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P143338 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08606940 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30778204 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30778195 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13008097 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12495359 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30778157 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30778234 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30778196 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30778205 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30778233 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30778151 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30778199 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30778146 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30778238 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30778202 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30778158 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30778143 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30778165 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30778229 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30778240 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30778141 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30778155 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30778160 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30778162 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30778226 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12785573 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30778231 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12803050 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30778237 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30778156 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13036366 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30778227 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30778242 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10439956 - -
Affymetrix Mouse Gene 1.0 ST Array 10439959 - -
Affymetrix Mouse Gene 1.0 ST Array 10439957 - -
Affymetrix HT MG-430 PM Array Plate 1445354_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1436941_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10439955 - -
Mouse Genome 430 2.0 Array 1436941_PM_at - -
Mouse Genome 430 2.0 Array 1445354_PM_at - -
Mouse Genome 430 2.0 Array 1436941_at - -
Mouse Genome 430 2.0 Array 1445354_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 385658_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 385658_PM_at - -
Affymetrix Mouse Clariom S Array PSR1600014094.mm.1 NM_001134457.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600014094.mm.1 NM_001134457.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600014094.mm.1 TC1600001743.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600014095.mm.1 NM_001134457.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600014095.mm.1 NM_001134457.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600014095.mm.1 TC1600001743.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600014093.mm.1 NM_001134457.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600014093.mm.1 NM_001134457.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600014093.mm.1 TC1600001743.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600014096.mm.1 NM_001134457.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600014096.mm.1 NM_001134457.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600014096.mm.1 TC1600001743.mm.1 MTA TC
Mouse Expression Set 430 1436941_PM_at - -
Mouse Expression Set 430 1445354_PM_at - -
Mouse Expression Set 430 1445354_at - -
Mouse Expression Set 430 1436941_at - -
Murine Genome U74v2 Set 166199_PM_at - -
Murine Genome U74v2 Set 166199_at - -
Affymetrix HT Mouse Genome 430B Array 1445354_at - -
Affymetrix HT Mouse Genome 430B Array 1445354_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1436941_at - -
Affymetrix HT Mouse Genome 430B Array 1436941_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 369020 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.1 expression beadchip ILMN_1251375 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2529953 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0