This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01147322_g1
See other SLTM GE Assays ›
Gene Symbol:
Sltm
Gene Name:
SAFB-like, transcription modulator
Gene Aliases:
5730455C01Rik, 5730555F13Rik, 9130215G10Rik, Met
Chromosome Location:
Chr.9: 70541796 - 70592237 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01147322_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sltm
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025690.3 NP_079966.2 11 - 12 1529 153
NM_026337.1 NP_080613.1 11 - 12 1567 153
GenBank mRNA AF462146.1 - 11 - 12 1529 153
AK017839.1 - 11 - 12 1428 153
AK045723.1 - 5 - 6 629 153
AK134756.1 - 2 - 3 208 153
BC139202.1 - 11 - 12 1519 8862224 153
BC145577.1 - 11 - 12 1519 153

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900000938.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sltm

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
biological_process
nucleotide binding
nucleic acid binding
RNA binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900007238.mm.1 NM_025690.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900007238.mm.1 NM_025690.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900007238.mm.1 NM_026337.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900007238.mm.1 NM_026337.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900007238.mm.1 TC0900000938.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0