This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01147328_m1
See other SLTM GE Assays ›
Gene Symbol:
Sltm
Gene Name:
SAFB-like, transcription modulator
Gene Aliases:
5730455C01Rik, 5730555F13Rik, 9130215G10Rik, Met
Chromosome Location:
Chr.9: 70541796 - 70592237 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01147328_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sltm
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025690.3 NP_079966.2 1 - 2 209 85
NM_026337.1 NP_080613.1 1 - 2 301 85
GenBank mRNA AF462146.1 - 1 - 2 209 85
AK011815.1 - 1 - 2 264 85
AK017839.1 - 1 - 2 162 85
AK077578.1 - 1 - 2 324 85
AK078251.1 - 1 - 2 286 85
AK162780.1 - 1 - 2 182 85
BC139202.1 - 1 - 2 199 8862224 85
BC145577.1 - 1 - 2 199 85

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900000938.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sltm

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
biological_process
nucleotide binding
nucleic acid binding
RNA binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900007220.mm.1 NM_025690.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900007220.mm.1 NM_025690.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900007220.mm.1 NM_026337.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900007220.mm.1 NM_026337.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900007220.mm.1 TC0900000938.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900007219.mm.1 NM_025690.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900007219.mm.1 NM_025690.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900007219.mm.1 NM_026337.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900007219.mm.1 NM_026337.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900007219.mm.1 TC0900000938.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900007216.mm.1 NM_026337.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900007216.mm.1 NM_026337.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900007216.mm.1 TC0900000938.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900007218.mm.1 NM_025690.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900007218.mm.1 NM_025690.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900007218.mm.1 NM_026337.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900007218.mm.1 NM_026337.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900007218.mm.1 TC0900000938.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 750284 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0