This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01149200_m1
See other YIF1B GE Assays ›
Gene Symbol:
Yif1b
Gene Name:
Yip1 interacting factor homolog B (S. cerevisiae)
Gene Aliases:
9430029K10Rik, Yip1b
Chromosome Location:
Chr.7: 29238308 - 29247532 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01149200_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Yif1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001110201.1 NP_001103671.1 5 - 6 590 105
NM_029887.3 NP_084163.2 5 - 6 599 105
GenBank mRNA AK135529.1 - 568 105
BC109331.2 - 5 - 6 569 40051521 105
BU921944.1 - 5 - 6 521 105
BU953116.1 - 5 - 6 587 105

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700000526.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Yif1b

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700004482.mm.1 TC0700000526.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700004482.mm.1 NM_001110201.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700004482.mm.1 NM_001110201.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700004482.mm.1 NM_029887.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700004482.mm.1 NM_029887.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0