This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01149202_m1
See other YIF1B GE Assays ›
Gene Symbol:
Yif1b
Gene Name:
Yip1 interacting factor homolog B (S. cerevisiae)
Gene Aliases:
9430029K10Rik, Yip1b
Chromosome Location:
Chr.7: 29238308 - 29247532 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01149202_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Yif1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001110201.1 NP_001103671.1 7 - 8 840 89
NM_029887.3 NP_084163.2 7 - 8 849 89
GenBank mRNA AK020444.1 - 7 - 8 820 89
AW743704.1 - 1 - 2 89
BC109331.2 - 7 - 8 819 40051521 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700000526.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Yif1b

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25026077 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25026137 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25026079 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25026136 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25026135 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25026080 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25026078 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2113774 BC109331.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2113774 NM_029887.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2113774 AK020444.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2113774 AK135529.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2113774 NM_001110201.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2102322 NM_029887.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2102322 AK020444.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2102322 AK135529.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2102322 AW743704.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2102322 NM_001110201.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 824850 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 654624 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 132447 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 3660 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 535187 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 504035 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 142595 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102141 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118602 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2102322 NM_029887.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2102322 AK020444.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2102322 AK135529.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2102322 AW743704.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2102322 NM_001110201.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2113774 BC109331.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2113774 NM_029887.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2113774 AK020444.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2113774 AK135529.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2113774 NM_001110201.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25026086 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25026069 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25026076 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25026078 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25026089 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25026067 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25026090 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25026071 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25026084 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25026088 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25026082 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25026079 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25026085 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25026074 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25026080 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P118602 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01491359 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P102141 - -
Whole Mouse Genome G4122A A_51_P256842 NM_029887.3 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P256842 AK020444.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P256842 AK135529.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P256842 AW743704.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P256842 NM_001110201.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P102141 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P118602 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06435120 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25026080 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25026078 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25026137 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25026079 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25026077 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25026136 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25026135 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026125 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026064 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026039 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026130 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026059 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026166 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026088 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026033 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026034 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026112 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026092 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026040 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026185 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026164 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026148 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026137 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026133 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026171 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026068 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026061 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026097 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026117 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026070 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026069 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026096 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026084 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026057 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026089 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026132 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026100 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026123 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026140 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026159 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026180 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026050 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026152 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026115 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026060 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026110 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026126 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026099 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026177 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026154 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026054 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026094 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026086 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026056 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026120 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026139 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026176 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026174 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026105 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026138 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026042 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026043 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026135 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026113 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026150 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026076 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026161 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026047 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026103 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026155 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026128 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026055 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026142 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026156 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026082 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026108 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026129 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026162 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026074 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026145 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026157 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026172 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026080 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25026037 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06435115 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026094 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06325066 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026084 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026126 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05887989 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026096 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06325067 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06391493 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026130 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026097 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026110 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026128 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06631254 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026131 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026092 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026121 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06181925 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06435122 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026076 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026071 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026088 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026137 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06391498 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026091 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026072 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05918263 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06396330 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026066 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026125 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026080 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026132 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05918256 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026069 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026117 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026135 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026068 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026067 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026089 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026100 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026112 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026105 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06391491 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06398413 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06435119 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026103 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25026108 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10551732 - -
Affymetrix Mouse Gene 1.0 ST Array 10551731 - -
Affymetrix Mouse Gene 1.0 ST Array 10551729 - -
Affymetrix Mouse Gene 1.0 ST Array 10551730 - -
Affymetrix Mouse Gene 1.0 ST Array 10551727 - -
Affymetrix Mouse Gene 1.0 ST Array 10551733 - -
Affymetrix Mouse Gene 1.0 ST Array 10551728 - -
Affymetrix Mouse Gene 1.0 ST Array 10551726 - -
Affymetrix Mouse Gene 1.0 ST Array 10551725 - -
Affymetrix HT MG-430 PM Array Plate 1428445_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10551724 - -
Affymetrix Mouse Gene 1.1 ST Array 10561706 - -
Mouse Genome 430 2.0 Array 1428445_PM_at - -
Mouse Genome 430 2.0 Array 1428445_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 77254_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 77254_PM_at - -
Affymetrix Mouse Clariom S Array PSR0700004485.mm.1 TC0700000526.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700004485.mm.1 NM_001110201.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700004485.mm.1 NM_001110201.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700004485.mm.1 NM_029887.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700004485.mm.1 NM_029887.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700004484.mm.1 TC0700000526.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700004484.mm.1 NM_001110201.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700004484.mm.1 NM_001110201.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700004484.mm.1 NM_029887.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700004484.mm.1 NM_029887.3 Refseq
Mouse Expression Set 430 1428445_PM_at - -
Mouse Expression Set 430 1428445_at - -
Murine Genome U74v2 Set 169001_f_at - -
Murine Genome U74v2 Set 169001_PM_f_at - -
Murine Genome U74v2 Set 171231_i_at - -
Murine Genome U74v2 Set 171231_PM_i_at - -
Affymetrix HT Mouse Genome 430B Array 1428445_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1428445_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 648975 - -
Array Name Probe IDs Targets Target Type
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2717304 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2607609 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2717304 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2607609 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0