This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01149627_m1
See other SMARCB1 GE Assays ›
Gene Symbol:
Smarcb1
Gene Name:
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1
Gene Aliases:
AU020204, Baf47, Ini1, SNF5/INI1, Snf5
Chromosome Location:
Chr.10: 75896769 - 75921614 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01149627_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Smarcb1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001161853.1 NP_001155325.1 1 - 2 313 88
NM_011418.2 NP_035548.1 1 - 2 313 88
GenBank mRNA AB017343.1 - 1 - 2 94 88
AB017344.1 - 1 - 2 94 88
AB041578.1 - 1 - 2 268 88
AJ011739.1 - 1 - 2 287 88
AJ011740.1 - 1 - 2 287 88
AK075906.1 - 1 - 2 296 88
AK145552.1 - 1 - 2 302 88
AK150169.1 - 1 - 2 314 88
AK185069.1 - 156 88
BC025163.1 - 1 - 2 267 3154501 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000002391.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Smarcb1

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

blastocyst development
blastocyst hatching
DNA repair
nucleosome disassembly
chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
cell cycle
nervous system development
negative regulation of cell proliferation
chromatin modification
cell differentiation
single stranded viral RNA replication via double stranded DNA intermediate
ATP-dependent chromatin remodeling
positive regulation by host of viral transcription
mitotic cell cycle phase transition
positive regulation of transcription from RNA polymerase II promoter
positive regulation of sequence-specific DNA binding transcription factor activity
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
p53 binding
transcription coactivator activity
protein binding
Tat protein binding
nucleosomal DNA binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000017120.mm.1 NM_001161853.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000017120.mm.1 NM_001161853.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000017120.mm.1 NM_011418.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000017120.mm.1 NM_011418.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000017120.mm.1 TC1000002391.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000017119.mm.1 NM_001161853.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000017119.mm.1 NM_001161853.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000017119.mm.1 NM_011418.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000017119.mm.1 NM_011418.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000017119.mm.1 TC1000002391.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000017122.mm.1 NM_001161853.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000017122.mm.1 NM_001161853.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000017122.mm.1 NM_011418.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000017122.mm.1 NM_011418.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000017122.mm.1 TC1000002391.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 889629 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-xql92:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline