Assay Details
| Gene Symbol: Elavl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010487.2 | NP_034617.1 | 3 - 4 | 760 | 66 | |
| GenBank mRNA | AK090281.1 | - | 3 - 4 | 760 | 66 | |
| BC052097.1 | - | 3 - 4 | 747 | 5695543 | 66 | |
| U29148.1 | - | 3 - 4 | 739 | 66 | ||
| U29149.1 | - | 1 - 2 | 80 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Elavl3
Genomic Map