This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01151963_gH
See other ELAVL3 GE Assays ›
Gene Symbol:
Elavl3
Gene Name:
ELAV (embryonic lethal, abnormal vision, Drosophila)-like 3 (Hu antigen C)
Gene Aliases:
2600009P04Rik, Huc, PLE21, mHuC
Chromosome Location:
Chr.9: 22015005 - 22052023 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01151963_gH
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Elavl3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010487.2 NP_034617.1 4 - 5 914 81
GenBank mRNA AK090281.1 - 4 - 5 914 81
BC052097.1 - 4 - 5 901 5695543 81
U29148.1 - 4 - 5 893 81
U29149.1 - 2 - 3 234 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900001892.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Elavl3

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
nervous system development
cell differentiation
nucleotide binding
nucleic acid binding
RNA binding
AU-rich element binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900015973.mm.1 NM_010487.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900015973.mm.1 NM_010487.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900015973.mm.1 TC0900001892.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0