This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01152074_m1
See other FAM214A GE Assays ›
Gene Symbol:
Fam214a
Gene Name:
family with sequence similarity 214, member A
Gene Aliases:
6330415I01Rik, AI553555, C130047D21Rik, Kiaa1370, mKIAA1370
Chromosome Location:
Chr.9: 74952919 - 75032468 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01152074_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fam214a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001113283.1 NP_001106754.1 5 - 6 863 63
NM_153584.2 NP_705812.2 4 - 5 686 63
GenBank mRNA AK173161.1 - 5 - 6 734 63
BC031353.1 - 4 - 5 686 5363230 63
BC060215.1 - 5 - 6 863 63
BU057452.1 - 4 - 5 600 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900001005.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fam214a

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900007812.mm.1 NM_001113283.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900007812.mm.1 NM_001113283.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900007812.mm.1 NM_153584.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900007812.mm.1 NM_153584.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900007812.mm.1 TC0900001005.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline