This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01156575_m1
See other MSRB3 GE Assays ›
Gene Symbol:
Msrb3
Gene Name:
methionine sulfoxide reductase B3
Gene Aliases:
D430026P16Rik, I-8-23, MsrB2
Chromosome Location:
Chr.10: 120781098 - 120899304 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01156575_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Msrb3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177092.4 NP_796066.1 3 - 4 370 68
GenBank mRNA AK086975.1 - 3 - 4 337 68
AK137163.1 - 3 - 4 337 68
BB617432.1 - 3 - 4 405 68
BC096447.1 - 3 - 4 251 6310686 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000003032.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Msrb3

Panther Classification:

Molecular Function -

oxidoreductase reductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

response to oxidative stress
protein repair
oxidation-reduction process
peptide-methionine (S)-S-oxide reductase activity
zinc ion binding
oxidoreductase activity
oxidoreductase activity, acting on a sulfur group of donors, disulfide as acceptor
peptide-methionine (R)-S-oxide reductase activity
metal ion binding
methionine-R-sulfoxide reductase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000022626.mm.1 NM_177092.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000022626.mm.1 NM_177092.4 Refseq
Affymetrix Mouse Clariom S Array PSR1000022626.mm.1 TC1000003032.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0