This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01156742_m1
See other CDH18 GE Assays ›
Gene Symbol:
Cdh18
Gene Name:
cadherin 18
Gene Aliases:
B230220E17Rik, Cdh14l
Chromosome Location:
Chr.15: 22548899 - 23474432 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01156742_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdh18
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081299.1 NP_001074768.1 5 - 6 1180 116
GenBank mRNA AK045672.1 - 5 - 6 1332 116
AK137430.1 - 5 - 6 1180 116

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500000142.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500001168.mm.1 NM_001081299.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500001168.mm.1 NM_001081299.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500001168.mm.1 TC1500000142.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0