This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01157590_m1
See other STARD4 GE Assays ›
Gene Symbol:
Stard4
Gene Name:
StAR-related lipid transfer (START) domain containing 4
Gene Aliases:
4632419C16Rik, 9030213J02Rik, AA517649, AW324468
Chromosome Location:
Chr.18: 33199140 - 33213879 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01157590_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stard4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133774.4 NP_598535.1 4 - 5 474 69
GenBank mRNA AF480297.1 - 4 - 5 329 69
AF480298.1 - 4 - 5 329 69
AK042347.1 - 4 - 5 478 69
AK054501.1 - 4 - 5 475 69
AK078858.1 - 4 - 5 478 69
AK085331.1 - 4 - 5 478 69
AK165641.1 - 4 - 5 480 69
AK204672.1 - 1 - 2 99 69
AK205539.1 - 1 - 2 112 69
BC005642.1 - 4 - 5 474 3709232 69

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stard4

Gene Ontology Categories:

Function(s) Process(es)

transport
lipid transport
lipid binding
cholesterol binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-g9ln6:80/100.66.78.28:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0