This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01157591_m1
See other STARD4 GE Assays ›
Gene Symbol:
Stard4
Gene Name:
StAR-related lipid transfer (START) domain containing 4
Gene Aliases:
4632419C16Rik, 9030213J02Rik, AA517649, AW324468
Chromosome Location:
Chr.18: 33199140 - 33213879 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01157591_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stard4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133774.4 NP_598535.1 5 - 6 584 89
GenBank mRNA AF480297.1 - 5 - 6 439 89
AF480298.1 - 5 - 6 439 89
AK042347.1 - 5 - 6 588 89
AK042691.1 - 5 - 6 544 89
AK054501.1 - 5 - 6 585 89
AK078858.1 - 5 - 6 588 89
AK085331.1 - 5 - 6 588 89
AK205539.1 - 2 - 3 222 89
BC005642.1 - 5 - 6 584 3709232 89

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stard4

Gene Ontology Categories:

Function(s) Process(es)

transport
lipid transport
lipid binding
cholesterol binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline