This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01157814_g1
See other TPGS2 GE Assays ›
Gene Symbol:
Tpgs2
Gene Name:
tubulin polyglutamylase complex subunit 2
Gene Aliases:
5730437P09Rik, 5730494M16Rik, AI666318, Pgs2
Chromosome Location:
Chr.18: 25127223 - 25169007 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01157814_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tpgs2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001004361.2 NP_001004361.2 3 - 4 523 77
NM_001142697.1 NP_001136169.1 3 - 4 269 77
NM_001142698.1 NP_001136170.1 3 - 4 523 77
GenBank mRNA AK017728.1 - 3 - 4 277 77
BC060253.1 - 1 - 2 75 77
BC080777.1 - 3 - 4 279 30439612 77
BQ770487.1 - 3 - 4 523 77
DV655125.1 - 3 - 4 269 77

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tpgs2

Panther Classification:

Molecular Function -

cytoskeletal protein microtubule or microtubule-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline