This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01158335_m1
See other CPED1 GE Assays ›
Gene Symbol:
Cped1
Gene Name:
cadherin-like and PC-esterase domain containing 1
Gene Aliases:
6720481P07, A430107O13Rik, AI552584
Chromosome Location:
Chr.6: 21985705 - 22256407 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01158335_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cped1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081351.1 NP_001074820.1 19 - 20 3527 102
GenBank mRNA AK032959.1 - 18 - 19 2214 102
BC151018.1 - 19 - 20 3042 9087931 102

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600000182.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600001197.mm.1 TC0600000182.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600001197.mm.1 NM_001081351.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600001197.mm.1 NM_001081351.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600001199.mm.1 TC0600000182.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600001199.mm.1 NM_001081351.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600001199.mm.1 NM_001081351.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600001196.mm.1 TC0600000182.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600001196.mm.1 NM_001081351.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600001196.mm.1 NM_001081351.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600001200.mm.1 TC0600000182.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600001200.mm.1 NM_001081351.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600001200.mm.1 NM_001081351.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0