| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600001181.mm.1 |
NM_001081351.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001181.mm.1 |
NM_001081351.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001181.mm.1 |
TC0600000182.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600001180.mm.1 |
NM_001081351.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001180.mm.1 |
NM_001081351.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001180.mm.1 |
TC0600000182.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600001182.mm.1 |
NM_001081351.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001182.mm.1 |
NM_001081351.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001182.mm.1 |
TC0600000182.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600001187.mm.1 |
NM_001081351.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001187.mm.1 |
NM_001081351.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001187.mm.1 |
TC0600000182.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600001185.mm.1 |
NM_001081351.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001185.mm.1 |
NM_001081351.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001185.mm.1 |
TC0600000182.mm.1 |
MTA TC |