This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01158493_g1
See other IK GE Assays ›
Gene Symbol:
Ik
Gene Name:
IK cytokine
Gene Aliases:
MuRED
Chromosome Location:
Chr.18: 36744656 - 36757639 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01158493_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011879.2 NP_036009.1 18 - 19 1724 65
GenBank mRNA AJ006130.1 - 18 - 19 1607 65
AK082356.1 - 18 - 19 1657 65
AK087976.1 - 18 - 19 1670 65
BC006706.1 - 16 - 17 2082 65
BC014739.1 - 18 - 19 1655 4920184 65
BC026569.1 - 18 - 19 1655 4982612 65
BC029148.1 - 16 - 17 2618 65
BC029671.1 - 18 - 19 1667 5364953 65
BC029694.1 - 18 - 19 1664 5369477 65
BC033356.1 - 18 - 19 1651 5366230 65
BM211698.2 - 2 - 3 65

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ik

Panther Classification:

Molecular Function -

cytokine

Gene Ontology Categories:

Process(es)

identical protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800002645.mm.1 NM_011879.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800002645.mm.1 NM_011879.2 Refseq
Affymetrix Mouse Clariom S Array PSR1800002645.mm.1 TC1800000343.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline