This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01158497_g1
See other IK GE Assays ›
Gene Symbol:
Ik
Gene Name:
IK cytokine
Gene Aliases:
MuRED
Chromosome Location:
Chr.18: 36744656 - 36757639 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01158497_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011879.2 NP_036009.1 6 - 7 640 70
GenBank mRNA AJ006130.1 - 6 - 7 523 70
AK029644.1 - 6 - 7 594 70
AK050625.1 - 6 - 7 598 70
AK082356.1 - 6 - 7 572 70
AK087976.1 - 6 - 7 586 70
AK163766.1 - 6 - 7 594 70
AK167238.1 - 6 - 7 605 70
BC014739.1 - 6 - 7 571 4920184 70
BC026569.1 - 6 - 7 571 4982612 70
BC029148.1 - 6 - 7 613 70
BC029671.1 - 6 - 7 583 5364953 70
BC029694.1 - 6 - 7 580 5369477 70
BC033356.1 - 6 - 7 567 5366230 70

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ik

Panther Classification:

Molecular Function -

cytokine

Gene Ontology Categories:

Process(es)

identical protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800002631.mm.1 NM_011879.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800002631.mm.1 NM_011879.2 Refseq
Affymetrix Mouse Clariom S Array PSR1800002631.mm.1 TC1800000343.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline