This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01158498_g1
See other IK GE Assays ›
Gene Symbol:
Ik
Gene Name:
IK cytokine
Gene Aliases:
MuRED
Chromosome Location:
Chr.18: 36744656 - 36757639 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01158498_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011879.2 NP_036009.1 7 - 8 707 89
GenBank mRNA AJ006130.1 - 7 - 8 590 89
AK029644.1 - 7 - 8 661 89
AK050625.1 - 7 - 8 665 89
AK082356.1 - 7 - 8 639 89
AK087976.1 - 7 - 8 653 89
AK088908.1 - 6 - 7 870 89
AK163766.1 - 7 - 8 661 89
AK167238.1 - 7 - 8 672 89
BC006706.1 - 5 - 6 1065 89
BC014739.1 - 7 - 8 638 4920184 89
BC026569.1 - 7 - 8 638 4982612 89
BC029148.1 - 7 - 8 680 89
BC029671.1 - 7 - 8 650 5364953 89
BC029694.1 - 7 - 8 647 5369477 89
BC033356.1 - 7 - 8 634 5366230 89

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ik

Panther Classification:

Molecular Function -

cytokine

Gene Ontology Categories:

Process(es)

identical protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800002632.mm.1 NM_011879.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800002632.mm.1 NM_011879.2 Refseq
Affymetrix Mouse Clariom S Array PSR1800002632.mm.1 TC1800000343.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline