This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01158499_m1
See other IK GE Assays ›
Gene Symbol:
Ik
Gene Name:
IK cytokine
Gene Aliases:
MuRED
Chromosome Location:
Chr.18: 36744656 - 36757639 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01158499_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011879.2 NP_036009.1 8 - 9 755 116
GenBank mRNA AJ006130.1 - 8 - 9 638 116
AK029644.1 - 8 - 9 709 116
AK050625.1 - 8 - 9 713 116
AK082356.1 - 8 - 9 687 116
AK087976.1 - 8 - 9 701 116
AK088908.1 - 7 - 8 918 116
AK163766.1 - 8 - 9 709 116
AK167238.1 - 8 - 9 720 116
BC006706.1 - 6 - 7 1113 116
BC014739.1 - 8 - 9 686 4920184 116
BC026569.1 - 8 - 9 686 4982612 116
BC029148.1 - 8 - 9 728 116
BC029671.1 - 8 - 9 698 5364953 116
BC029694.1 - 8 - 9 695 5369477 116
BC033356.1 - 8 - 9 682 5366230 116

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800000343.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ik

Panther Classification:

Molecular Function -

cytokine

Gene Ontology Categories:

Process(es)

identical protein binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800002633.mm.1 NM_011879.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800002633.mm.1 NM_011879.2 Refseq
Affymetrix Mouse Clariom S Array PSR1800002633.mm.1 TC1800000343.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 374711 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline