This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01158514_m1
See other APBB3 GE Assays ›
Gene Symbol:
Apbb3
Gene Name:
amyloid beta (A4) precursor protein-binding, family B, member 3
Gene Aliases:
Fe65l2, Rirl2, TR2S
Chromosome Location:
Chr.18: 36671154 - 36679366 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01158514_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Apbb3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_146085.1 NP_666197.1 12 - 13 1542 61
GenBank mRNA AK141878.1 - 12 - 13 1444 61
AK171634.1 - 11 - 12 1735 61
BC024136.1 - 11 - 12 1540 61
BC024809.1 - 12 - 13 1542 5357681 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800001748.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Apbb3

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
negative regulation of apoptotic process
beta-amyloid binding
transcription factor binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625259 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625258 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625255 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625256 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625260 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625257 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2049582 BC024136.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2049582 BC024809.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2049582 AK171634.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2049582 NM_146085.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 389010 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 242280 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 165261 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 819941 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 972549 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125778 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2049582 BC024136.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2049582 BC024809.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2049582 AK171634.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2049582 NM_146085.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P277321 BC024136.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P277321 BC024809.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P277321 AK171634.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P277321 AK141878.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P277321 NM_146085.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625248 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625253 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625234 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625255 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625193 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625241 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625197 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625247 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625229 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625192 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625221 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625239 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625243 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625256 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625214 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625237 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625194 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625235 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625236 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625238 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625215 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625257 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625223 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625219 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625233 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625226 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625252 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625196 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625245 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625260 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625224 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625240 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08966791 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125778 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08966807 - -
Whole Mouse Genome G4122A A_51_P277321 BC024136.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P277321 BC024809.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P277321 AK171634.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P277321 AK141878.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P277321 NM_146085.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114159 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125778 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13738784 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13738773 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13738779 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13738774 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625256 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625258 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625255 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625260 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625257 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625259 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625127 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625171 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625121 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625274 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625260 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625146 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625161 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625141 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625275 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625137 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625266 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625158 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625150 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625322 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625261 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625147 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625163 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625325 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625268 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625135 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625329 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625152 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625173 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625138 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625165 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625164 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625118 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625167 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625123 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625176 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625131 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625129 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625133 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625125 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625124 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625262 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625155 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625128 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625166 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625264 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625169 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625175 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625332 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13895734 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31625262 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31625274 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31625275 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31625261 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31625264 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14091979 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31625271 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31625260 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31625289 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31625293 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31625266 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31625268 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10458361 - -
Affymetrix Mouse Gene 1.0 ST Array 10458366 - -
Affymetrix Mouse Gene 1.0 ST Array 10458367 - -
Affymetrix Mouse Gene 1.0 ST Array 10458364 - -
Affymetrix Mouse Gene 1.0 ST Array 10458358 - -
Affymetrix Mouse Gene 1.0 ST Array 10458357 - -
Affymetrix Mouse Gene 1.0 ST Array 10458356 - -
Affymetrix Mouse Gene 1.0 ST Array 10458362 - -
Affymetrix Mouse Gene 1.0 ST Array 10458365 - -
Affymetrix Mouse Gene 1.0 ST Array 10458363 - -
Affymetrix Mouse Gene 1.0 ST Array 10458360 - -
Affymetrix Mouse Gene 1.0 ST Array 10458359 - -
Affymetrix HT MG-430 PM Array Plate 1424457_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10458355 - -
Mouse Genome 430 2.0 Array 1424457_at - -
Mouse Genome 430 2.0 Array 1424457_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 225372_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 225372_PM_at - -
Affymetrix Mouse Clariom S Array PSR1800012325.mm.1 NM_146085.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800012325.mm.1 NM_146085.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800012325.mm.1 TC1800001748.mm.1 MTA TC
Mouse Expression Set 430 1424457_at - -
Mouse Expression Set 430 1424457_PM_at - -
Mouse Genome 430A 2.0 Array 1424457_PM_at - -
Mouse Genome 430A 2.0 Array 1424457_at - -
Affymetrix HT Mouse Genome 430A Array 1424457_at - -
Affymetrix HT Mouse Genome 430A Array 1424457_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 722977 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 6020168 - -
Illumina mouseRef-8 v1.0 expression beadchip 0006020168 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1223271 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1223271 - -
mouse-6 v1.1 (Illumina) scl51509.11.1_166-S - -
Illumina Sentrix mouse-6.v1 bead array 6020168 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl51509.11.1_166-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 6020168 - -
MouseRef-8 v2 Expression BeadChip ILMN_1223271 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1223271 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl51509.11.1_166-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0